Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaric Dialdehyde, 50% in Water, Spectrum™ Chemical
SDP

Catalog No. 18610724 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-724 4 L Amber Glass Bottle
18-610-722 100 mL Amber Glass Bottle
18-610-723 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-610-724 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. G30604LT
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-30-8 , 7732-18-5

Specifications

Boiling Point 100.7°C to 101°C (213.3°F to 213.8°F)
Packaging Amber Glass Bottle
Quantity 4 L
Grade Reagent
Physical Form Liquid
Chemical Name or Material Glutaric Dialdehyde, 50 Percent in Water

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.