Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glutarimide-isoindolinone-NH-PEG4-COOH | 2927334-86-7 | 99.5% | 507.5 g·mol⁻¹ | C24H33N3O9 | 5 MG
SDP

Catalog No. 5000223429
Encompass_Preferred

Glutarimide-Isoindolinone-NH-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a cereblon-binding glutarimide-isoindolinone core with a PEG4 linker terminating in a carboxylic acid, intended for use in PROTAC research to recruit CRBN and promote ubiquitin-proteasome-mediated degradation of target proteins.

  • Designed for PROTAC development to recruit cereblon E3 ligase.
  • PEG4 linker with terminal carboxylic acid enables facile conjugation to target ligands.
  • High reported purity suitable for research applications.
  • Soluble in DMSO; published in vitro and in vivo formulation guidance available.
  • Store at -20°C and protect from light; stability guidance provided for solutions.

Catalog No. 50-002-23429 Supplier Medchemexpress LLC Supplier No. HY1387905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutarimide-Isoindolinone-NH-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that combines a cereblon-binding glutarimide-isoindolinone core with a PEG4 linker terminating in a carboxylic acid, intended for use in PROTAC research to recruit CRBN and promote ubiquitin-proteasome-mediated degradation of target proteins.

  • Designed for PROTAC development to recruit cereblon E3 ligase.
  • PEG4 linker with terminal carboxylic acid enables facile conjugation to target ligands.
  • High reported purity suitable for research applications.
  • Soluble in DMSO; published in vitro and in vivo formulation guidance available.
  • Store at -20°C and protect from light; stability guidance provided for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.