Learn More
Medchemexpress LLC Glutarimide-isoindolinone-NH-PEG4-COOH | 2927334-86-7 | 99.5% | 507.53 | 100 MG
SDP
Catalog No. 5000351205
Encompass_Preferred
Glutarimide-Isoindolinone-NH-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a cereblon ligand and a linker used in PROTAC technology. Proteolysis Targeting Chimeras (PROTACs) consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Used in PROTAC technology
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51205 Supplier Medchemexpress LLC Supplier No. HY138790100MG
Glutarimide-Isoindolinone-NH-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a cereblon ligand and a linker used in PROTAC technology. Proteolysis Targeting Chimeras (PROTACs) consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Used in PROTAC technology
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins