Learn More
Medchemexpress LLC Glutarimide-Isoindolinone-NH-PEG4-COOH | 2927334-86-7 | 99.5% | 507.53 | 50 MG
SDP
Catalog No. 5000348884
Encompass_Preferred
Glutarimide-Isoindolinone-NH-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a cereblon ligand and a linker used in PROTAC technology. It is intended for research use only.
  • This compound is designed to act as an E3 ligase ligand-linker conjugate.
  • It includes a cereblon ligand.
  • It contains a linker used in PROTAC technology.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • The product has a purity of 99.5%.
Catalog No. 50-003-48884 Supplier Medchemexpress LLC Supplier No. HY13879050MG
Glutarimide-Isoindolinone-NH-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a cereblon ligand and a linker used in PROTAC technology. It is intended for research use only.
  • This compound is designed to act as an E3 ligase ligand-linker conjugate.
  • It includes a cereblon ligand.
  • It contains a linker used in PROTAC technology.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • The product has a purity of 99.5%.