Learn More
Apexbio Technology LLC GLUTARYL-CARNITINE-D3-HCL-1MG
SDP
Catalog No. 5000570420
Encompass_Preferred
Glutaryl carnitine-d3 HCl is a stable isotope-labeled compound used predominantly in biomedical research particularly in studies related to metabolic disorders and mitochondrial function Its mechanism of action involves participation in fatty acid metabolism and energy production targeting pathways within the mitochondria This compound often serves as a standard or tracer in mass spectrometry to quantify levels of carnitine derivatives within biological samples In vitro studies have demonstrated activity within the nanomolar to low micromolar range It is commonly applied in research involving cell models such as hepatocytes and in animal studies to elucidate its role in metabolic pathways Additionally it serves as a critical tool in drug screening assays assisting in the evaluation of new therapeutic approaches for metabolic dysfunctions Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70420 Supplier Apexbio Technology LLC Supplier No. SL10251
Glutaryl carnitine-d3 HCl is a stable isotope-labeled compound used predominantly in biomedical research particularly in studies related to metabolic disorders and mitochondrial function Its mechanism of action involves participation in fatty acid metabolism and energy production targeting pathways within the mitochondria This compound often serves as a standard or tracer in mass spectrometry to quantify levels of carnitine derivatives within biological samples In vitro studies have demonstrated activity within the nanomolar to low micromolar range It is commonly applied in research involving cell models such as hepatocytes and in animal studies to elucidate its role in metabolic pathways Additionally it serves as a critical tool in drug screening assays assisting in the evaluation of new therapeutic approaches for metabolic dysfunctions Experimental concentrations typically depend on the specific research design