Learn More
Medchemexpress LLC Glutaryl coenzyme A lithium | 103192-48-9 | 99.75% | C26H40Li2N7O19P3S | 1 MG
SDP
Catalog No. 5000353432
Encompass_Preferred
Glutaryl coenzyme A lithium is an important endogenous metabolite used in HMG-CoA or Glutaryl-CoA related experiments. It is intended for research use only.
  • It is supplied as a solid that is white to off-white in color.
  • It can be dissolved in water at a concentration of 100 mg/mL.
  • Shipping is at room temperature in the continental US.
  • Storage is recommended at -20°C, sealed and away from moisture.
  • Stock solutions can be stored at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-53432 Supplier Medchemexpress LLC Supplier No. HY1414741MG
Glutaryl coenzyme A lithium is an important endogenous metabolite used in HMG-CoA or Glutaryl-CoA related experiments. It is intended for research use only.
  • It is supplied as a solid that is white to off-white in color.
  • It can be dissolved in water at a concentration of 100 mg/mL.
  • Shipping is at room temperature in the continental US.
  • Storage is recommended at -20°C, sealed and away from moisture.
  • Stock solutions can be stored at -80°C for 6 months or -20°C for 1 month.