Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glutarylcarnitine-d9 chloride | 95.0% | 320.81 g/mol | C12H13D9ClNO6 | 5 MG
SDP

Catalog No. 5000264289
Encompass_Preferred

Deuterium-labeled glutarylcarnitine chloride supplied as a 5 mg analytical standard for use as an internal standard in mass spectrometry and quantitative metabolite analysis. Provided as an oil with stated purity.

  • Deuterium-labeled internal standard for glutarylcarnitine.
  • Suitable for mass spectrometry and quantitative assays.
  • 5 mg package size for calibration and method development.
  • High purity (95.0%) to support accurate quantitation.
  • Chloride salt provided in oil form for stability.
  • Molecular weight 320.81 g/mol; formula C12H13D9ClNO6.

Catalog No. 50-002-64289 Supplier Medchemexpress LLC Supplier No. HY113005S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled glutarylcarnitine chloride supplied as a 5 mg analytical standard for use as an internal standard in mass spectrometry and quantitative metabolite analysis. Provided as an oil with stated purity.

  • Deuterium-labeled internal standard for glutarylcarnitine.
  • Suitable for mass spectrometry and quantitative assays.
  • 5 mg package size for calibration and method development.
  • High purity (95.0%) to support accurate quantitation.
  • Chloride salt provided in oil form for stability.
  • Molecular weight 320.81 g/mol; formula C12H13D9ClNO6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.