Learn More
Cayman Chemical Glutathione Assay Kit, 480 Well
SDP
Catalog No. 502893607
Encompass_Preferred
Cayman’s GSH Assay utilizes a carefully optimized enzymatic recycling method for the quantification of GSH in a 96-well microplate format. It measures both GSH and GSSG to reflect total glutathione in a sample. The kit can also be used to measure only GSSG by following an alternative protocol. The GSH Assay can be used for plasma, tissue samples, and cultured cells with minimal sample processing.
Catalog No. 50-289-3607 Supplier Cayman Chemical Supplier No. 703002480 WELL
Cayman’s GSH Assay utilizes a carefully optimized enzymatic recycling method for the quantification of GSH in a 96-well microplate format. It measures both GSH and GSSG to reflect total glutathione in a sample. The kit can also be used to measure only GSSG by following an alternative protocol. The GSH Assay can be used for plasma, tissue samples, and cultured cells with minimal sample processing.