Learn More
Cayman Chemical GlutathIon ethyl ester 50mg
SDP
Catalog No. 502972755
Encompass_Preferred
A cell-permeable derivative of GSH; increases mitochondrial GSH levels in C38 and IB3-1 epithelial lung cystic fibrosis cells at 10 mM; decreases reactive oxygen species (ROS) levels and TNF-α-induced IL-8 secretion in IB3-1 cells at 10 mM; increases mitochondrial complex I function and reduces depolarization of the mitochondrial membrane potential in IB3-1 cells at 10 mM and 1mM, respectively; decreases lipid oxidation and ferroptosis in cystine-deficient PANC-1, AsPC-1, BxPC-3, and S2-013 pancreatic cancer cells; increases the viability of irradiated human lymphoid cells at 5 mM
Catalog No. 50-297-2755 Supplier Cayman Chemical Supplier No. 1495350 MG
A cell-permeable derivative of GSH; increases mitochondrial GSH levels in C38 and IB3-1 epithelial lung cystic fibrosis cells at 10 mM; decreases reactive oxygen species (ROS) levels and TNF-α-induced IL-8 secretion in IB3-1 cells at 10 mM; increases mitochondrial complex I function and reduces depolarization of the mitochondrial membrane potential in IB3-1 cells at 10 mM and 1mM, respectively; decreases lipid oxidation and ferroptosis in cystine-deficient PANC-1, AsPC-1, BxPC-3, and S2-013 pancreatic cancer cells; increases the viability of irradiated human lymphoid cells at 5 mM