Learn More
Cayman Chemical Glutathione Reductase Assay Kit, 96 Well
SDP
Catalog No. 502893445
Encompass_Preferred
GR is a flavoprotein that catalyzes the NADPH-dependent reduction of oxidized GSSG to GSH. This enzyme is essential for the GSH redox cycle which maintains adequate levels of reduced cellular GSH. A high GSH/GSSG ratio is essential for protection against oxidative stress. Cayman’s Glutathione Reductase Assay Kit measures GR activity by measuring the rate of NADPH oxidation. The oxidation of NADPH to NADP+ is accompanied by a decrease in absorbance at 340 nm. The rate of decrease in the absorbance at 340 nm is directly proportional to the GR activity in the sample.
Catalog No. 50-289-3445 Supplier Cayman Chemical Supplier No. 70320296 WELL
GR is a flavoprotein that catalyzes the NADPH-dependent reduction of oxidized GSSG to GSH. This enzyme is essential for the GSH redox cycle which maintains adequate levels of reduced cellular GSH. A high GSH/GSSG ratio is essential for protection against oxidative stress. Cayman’s Glutathione Reductase Assay Kit measures GR activity by measuring the rate of NADPH oxidation. The oxidation of NADPH to NADP+ is accompanied by a decrease in absorbance at 340 nm. The rate of decrease in the absorbance at 340 nm is directly proportional to the GR activity in the sample.