Learn More
Ubpbio GLUTATHIONE AGAROSE, 10ML
Catalog No. 501126952
Encompass_Preferred
Protein Purification Reagents – Glutathione Resin (UBPBio catalog# P3060-10, P3060-100) – Glutathione Xpure Agarose Resin consists of glutathione covalently bound to agarose beads. This resin has high binding affinity and specificity to the Glutathione S-transferase (GST) protein. GST-tagged proteins can be highly purified by batch or column purification (low flow rate) in a one step purification method. Bound GST-tagged proteins are easily eluted from the resin with buffers containing reduced glutathione.
Catalog No. 50-112-6952 Supplier Ubpbio Supplier No. P306010
Protein Purification Reagents – Glutathione Resin (UBPBio catalog# P3060-10, P3060-100) – Glutathione Xpure Agarose Resin consists of glutathione covalently bound to agarose beads. This resin has high binding affinity and specificity to the Glutathione S-transferase (GST) protein. GST-tagged proteins can be highly purified by batch or column purification (low flow rate) in a one step purification method. Bound GST-tagged proteins are easily eluted from the resin with buffers containing reduced glutathione.