Learn More
Gold Biotechnology Inc Glutathione Agarose Resin 50 mL
SDP
Catalog No. 501532779
Encompass_Preferred
Glutathione has been covalently linked for use in affinity purification of glutathione-S-transferase (GST) and GST fusion proteins.
Catalog No. 50-153-2779 Supplier Gold Biotechnology Inc Supplier No. G25050
Glutathione has been covalently linked for use in affinity purification of glutathione-S-transferase (GST) and GST fusion proteins.