Learn More
Gold Biotechnology Inc Glutathione Agarose Resin 100 mL
SDP
Catalog No. 501532777
Encompass_Preferred
Glutathione has been covalently linked for use in affinity purification of glutathione-S-transferase (GST) and GST fusion proteins.
Catalog No. 50-153-2777 Supplier Gold Biotechnology Inc Supplier No. G250100
Glutathione has been covalently linked for use in affinity purification of glutathione-S-transferase (GST) and GST fusion proteins.