Learn More
TARGETMOL CHEMICALS INC GLUTATHIONE ARSENOXIDE H 5MG
SDP
Catalog No. 502580615
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Glutathione arsenoxide hydrochloride (Glutathione arsenoxide hydrochloride(1271726-51-2 Free base)) is a potentially active anti-cancer molecule and inhibitor of tumour metabolism. glutathione arsenoxide hydrochloride targets the mitochondrial inner membrane adenine nucleotidyl transferase (ANT) which inhibits cell proliferation and promotes apoptosis. Glutathione arsenoxide hydrochloride can be used to recognize cell surface proteins such as protein disulfide isomerases.
Catalog No. 50-258-0615 Supplier TARGETMOL CHEMICALS INC Supplier No. T27417L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Glutathione arsenoxide hydrochloride (Glutathione arsenoxide hydrochloride(1271726-51-2 Free base)) is a potentially active anti-cancer molecule and inhibitor of tumour metabolism. glutathione arsenoxide hydrochloride targets the mitochondrial inner membrane adenine nucleotidyl transferase (ANT) which inhibits cell proliferation and promotes apoptosis. Glutathione arsenoxide hydrochloride can be used to recognize cell surface proteins such as protein disulfide isomerases.