Learn More
XpressBio Xpress Capture Glutathione ELISA Plates, Clear 5x96 wells (5-pack)
SDP
Supplier:  XpressBio EX7000C96
Encompass_Preferred
Capture coated Glutathione ELISA plates may be used with GST-tagged proteins for several uses including recombinant protein expression screening, immunoabsorbtion assays, biochemical assays, competition assays and protein purification.
Catalog No. 50-171-7646