Learn More
Bioworld Glutathione-Coated Microplates, 5 Plate(s)
Catalog No. 50197550
Encompass
bioWorld's Glutathione Coated Plates are standard 96-well polystyrene microplates whosesurface has been coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins. Applications:Analysis of cell lysates to determine the presence and concentration of the GST-fusion protein produced in an expression system. Alternatively, the plates can be used to capture and coat fusion proteins via their GST tag for subsequent binding assays involving the recombinant protein (either protein interactions or a specific antibody). Because the plates are pre-blocked, initial purification of the cell lysates is not necessary.
Catalog No. 50-197-550 Supplier Bioworld Supplier No. 201400191
bioWorld's Glutathione Coated Plates are standard 96-well polystyrene microplates whosesurface has been coated with intact glutathione (GSH) and pre-blocked for use in capture and detection assays involving glutathione-S-transferase (GST) fusion proteins. Applications:Analysis of cell lysates to determine the presence and concentration of the GST-fusion protein produced in an expression system. Alternatively, the plates can be used to capture and coat fusion proteins via their GST tag for subsequent binding assays involving the recombinant protein (either protein interactions or a specific antibody). Because the plates are pre-blocked, initial purification of the cell lysates is not necessary.