Learn More
Arbor Assays Glutathione Fluorescent Detection Kit (5 Plate)
SDP
Catalog No. 502278385
Encompass_Preferred
The DetectX Glutathione (GSH) Fluorescent Detection Kit utilizes a proprietary non-fluorescent molecule, ThioStar, that covalently binds to the free thiol group on GSH to yield a highly fluorescent product.
This item is not returnable. View return policy
Catalog No. 50-227-8385 Supplier Arbor Assays Supplier No. K006F5
Add to Cart
Add to Cart
The DetectX Glutathione (GSH) Fluorescent Detection Kit utilizes a proprietary non-fluorescent molecule, ThioStar, that covalently binds to the free thiol group on GSH to yield a highly fluorescent product.