Learn More
Medchemexpress LLC Glutathione diethyl ester (TFA) | 97.35% | 477.45 | 1 MG
SDP
Catalog No. 5000398510
Encompass_Preferred
Glutathione diethyl ester (TFA) is a delivery agent for glutathione monoester, and consequently for glutathione, in human cells. It can potentially reduce oxidative stress and toxicity. This product is for research use only and is not sold to patients.
  • Delivery agent for glutathione monoester
  • May decrease oxidative stress
  • May decrease toxicity
  • Classified as an endogenous metabolite
  • Relevant to metabolic and neurodegenerative disease research
  • Also used in neurological and eye or ear disease research
  • A peptide derivative and natural product standard
Catalog No. 50-003-98510 Supplier Medchemexpress LLC Supplier No. HYP5069A1MG
Glutathione diethyl ester (TFA) is a delivery agent for glutathione monoester, and consequently for glutathione, in human cells. It can potentially reduce oxidative stress and toxicity. This product is for research use only and is not sold to patients.
  • Delivery agent for glutathione monoester
  • May decrease oxidative stress
  • May decrease toxicity
  • Classified as an endogenous metabolite
  • Relevant to metabolic and neurodegenerative disease research
  • Also used in neurological and eye or ear disease research
  • A peptide derivative and natural product standard