Learn More
Medchemexpress LLC Glutathione ethyl ester | 92614-59-0 | MFCD00156117 | 95.8% | 335.38 g·mol⁻¹ | C12H21N3O6S | 5 MG
SDP
Catalog No. 5000262703
Encompass_Preferred
Glutathione ethyl ester is a cell-permeable glutathione (GSH) donor used to raise intracellular GSH levels and enhance antioxidative defenses in cell culture and reproductive biology applications.
  • Cell-permeable glutathione donor
  • Increases intracellular GSH levels
  • Enhances antioxidative defense in cultured cells and oocytes
  • Supports in vitro embryo and oocyte culture applications
  • Supplied as a solid powder for dissolution into media
  • Purity 95.8% as listed
Catalog No. 50-002-62703 Supplier Medchemexpress LLC Supplier No. HY1341245MG
Glutathione ethyl ester is a cell-permeable glutathione (GSH) donor used to raise intracellular GSH levels and enhance antioxidative defenses in cell culture and reproductive biology applications.
  • Cell-permeable glutathione donor
  • Increases intracellular GSH levels
  • Enhances antioxidative defense in cultured cells and oocytes
  • Supports in vitro embryo and oocyte culture applications
  • Supplied as a solid powder for dissolution into media
  • Purity 95.8% as listed