Learn More
Medchemexpress LLC Glutathione ethyl ester | 92614-59-0 | MFCD00156117 | 95.8% | C12H21N3O6S | 10MG
SDP
Catalog No. 5000261383
Encompass_Preferred
Glutathione ethyl ester is an esterified, cell-permeable derivative of glutathione that supplies intracellular GSH. It functions as an efficient GSH donor to restore cellular antioxidant capacity and is used in cell and embryo culture to protect against oxidative stress.
  • Cell-permeable glutathione donor that increases intracellular GSH levels.
  • Enhances antioxidative defense in oocytes and embryos.
  • Suitable for cell culture and biochemical research applications.
  • Available as solid powder or DMSO solution for convenient reconstitution.
  • Reported purity approximately 95.8%.
Catalog No. 50-002-61383 Supplier Medchemexpress LLC Supplier No. HY13412410MG
Add to Cart
Add to Cart
Glutathione ethyl ester is an esterified, cell-permeable derivative of glutathione that supplies intracellular GSH. It functions as an efficient GSH donor to restore cellular antioxidant capacity and is used in cell and embryo culture to protect against oxidative stress.
  • Cell-permeable glutathione donor that increases intracellular GSH levels.
  • Enhances antioxidative defense in oocytes and embryos.
  • Suitable for cell culture and biochemical research applications.
  • Available as solid powder or DMSO solution for convenient reconstitution.
  • Reported purity approximately 95.8%.