Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutathione Fluorescent Detection Kit

Catalog No. EIAGSHF
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAGSHF 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAGSHF Supplier Invitrogen™ Supplier No. EIAGSHF
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

96-well plates, standard, other assay specific reagents, buffers and lot-specific technical data sheet.

Assay Glutathione levels in Whole Blood, Serum, Plasma, Erythrocytes, Urine, Cell Lysates and Tissue Samples (fluorescent).

The Glutathione Detection research-use-only kit is a fluorescent assay designed for the quantification and detection of glutathione in whole blood, serum, plasma, erythrocytes, urine, cell lysates and tissue samples.

This complete, ready-to-use kit includes black 96-well plate(s), glutathione standard, detection reagent, assay buffer, and other components to perform the assay. A 96-well microplate reader capable of reading fluorescent emission at 510 nm, with excitation at 390 nm, is required for use of this kit.

Performance characteristics
• Assay type: fluorescent detection kit
• Sample types: whole blood, serum, plasma, erythrocytes, urine, cell lysates, and tissue samples
• Sensitivity: 45 nM
• Standard curve range: 0.2–25 μM
• Reactivity: species independent

Background
Glutathione (GSH) is the highest concentration non-protein thiol in mammalian cells and is present in concentrations of 0.5–10 mM. GSH plays a key role in many biological processes, including the synthesis of proteins and DNA, the transport of amino acids, and the protection of cells against oxidation. Harmful hydrogen peroxide cellular levels are minimized by the enzyme glutathione peroxidase (GP) using GSH as a reductant. The oxidized GSH dimer, GSSG, is formed from GSH and peroxide by the GP reaction.

An important role of GSSG in the NFkappaB activating signal cascade is suggested by the facts that the potent NFkB inducer, tetradecanoyl phorbol acetate, increases intracellular GSSG levels and GSSG/GSH ratios. Glutathione S-transferases (GST) are an important group of enzymes that catalyze the nucleophilic addition of GSH to electrophiles. They are encoded by 5 gene families; 4 encode cytosolic GST and one encodes the microsomal form of GST.

Assay principle
The Glutathione Fluorescent Detection Kit is designed to quantitatively measure glutathione (GSH), and oxidized glutathione (GSSG) present in a variety of samples. The kit is unique in that both free and oxidized glutathione are detected in the same well in the microtiter plate. No separation or washing is required. Total glutathione is the sum of GSSG plus GSH. A GSH standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. The kit utilizes a proprietary non-fluorescent detection reagent, that will covalently bind to the free thiol group on GSH to yield a highly fluorescent product. After mixing the sample or standard with the detection reagent and incubating at room temperature for 15 minutes, the fluorescent product is read at 510 nm in a fluorescent plate reader with excitation at 390 nm. Free glutathione, GSH, is read first after 15 minutes, followed by addition of a reaction mixture that converts all the oxidized glutathione, GSSG, into free GSH, which then reacts with the excess detection reagent to yield the signal related to Total GSH content. The total concentration of GSH generated in the sample is calculated from the generated signal.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at –20°C.
Immunoassay Kit Format Fluorescent Detection Kit
Product Type Glutathione Fluorescent Detection Kit
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Quantity 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Glutathione

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.