Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutathione High Capacity
Catalog No. 502904861
Encompass
Glutathione Magnetic Agarose Beads consist of a paramagnetic 4% beaded agarose. Glutathione is covalently attached through the sulfur to epoxy-activatedagarose resulting in a 12 atom (10 carbon) spacer.The Glutathione Magnetic Agarose Beads are useful for affinity capture molecular pull-down or immunoprecipitation (IP) of proteins containing glutathione binding sequences such as native glutathione S-transferase (GST) glutathione peroxidase and glyoxalase I or GST-tagged recombinant fusion proteins from cell lysates or other biochemical SOLUTIONons while exhibiting low non-specific binding of other proteins. Recombinant GST-tagged proteins bound to the affinity resin are separated with the use of a magnet. The magnetic properties allow for very rapid processing as well as aid in manipulations such as repetitive washings and recovery of the desired protein. This leads to faster recovery experimental reproducibility and more accurate quantitation of the proteins of interest.The
Catalog No. 50-290-4861 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G09245X1ML
Glutathione Magnetic Agarose Beads consist of a paramagnetic 4% beaded agarose. Glutathione is covalently attached through the sulfur to epoxy-activatedagarose resulting in a 12 atom (10 carbon) spacer.The Glutathione Magnetic Agarose Beads are useful for affinity capture molecular pull-down or immunoprecipitation (IP) of proteins containing glutathione binding sequences such as native glutathione S-transferase (GST) glutathione peroxidase and glyoxalase I or GST-tagged recombinant fusion proteins from cell lysates or other biochemical SOLUTIONons while exhibiting low non-specific binding of other proteins. Recombinant GST-tagged proteins bound to the affinity resin are separated with the use of a magnet. The magnetic properties allow for very rapid processing as well as aid in manipulations such as repetitive washings and recovery of the desired protein. This leads to faster recovery experimental reproducibility and more accurate quantitation of the proteins of interest.The