Learn More
Medchemexpress LLC Glutathione oxidized | 27025-41-8 | 98.46% | 250 MG
SDP
Catalog No. 5000413746
Encompass_Preferred
Glutathione oxidized (Standard) serves as the analytical standard for Glutathione oxidized, designated for research and analytical applications. It is formed through the oxidation of glutathione, a process that occurs during the detoxification of reactive oxygen species. This product can be utilized for research pertaining to sickle cells and erythrocytes.
  • It is the analytical standard of glutathione oxidized.
  • Intended for research and analytical applications.
  • Produced by the oxidation of glutathione.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-13746 Supplier Medchemexpress LLC Supplier No. 250MGHYD0844R
Glutathione oxidized (Standard) serves as the analytical standard for Glutathione oxidized, designated for research and analytical applications. It is formed through the oxidation of glutathione, a process that occurs during the detoxification of reactive oxygen species. This product can be utilized for research pertaining to sickle cells and erythrocytes.
  • It is the analytical standard of glutathione oxidized.
  • Intended for research and analytical applications.
  • Produced by the oxidation of glutathione.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.