Learn More
Enzo Life Sciences Glutathione peroxidase activity kit (5x96wells)
SDP
Catalog No. 50658184
Encompass_Preferred
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes. Alternative Name: GPx. Applications: Colorimetric detection, Activity assay. Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG
Catalog No. 50-658-184 Supplier Enzo Life Sciences Supplier No. ADI900158
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes. Alternative Name: GPx. Applications: Colorimetric detection, Activity assay. Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG