Learn More
Biotage GLUTATHIONE PHYTIP COL 300 UL
SDP
Catalog No. NC3312119
Encompass
Glutathione PhyTip® columns (300 µL volume) containing 20 µL affinity media per column, DD/Biotix 96pk
Catalog No. NC3312119 Supplier Biotage Supplier No. PTX932004
Glutathione PhyTip® columns (300 µL volume) containing 20 µL affinity media per column, DD/Biotix 96pk