Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione, Reduced, Immobilized on Agarose CL-4B, For affinity chromatography, Powder (lyophilized), MilliporeSigma™ Supelco™

Catalog No. 111003043 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3043 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3043 Supplier MilliporeSigma Supelco Supplier No. 4973950ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

the lactose stabilizer must be removed prior to use by washing the gel onto filter with water or buffer; the reduced glutathione is attached through the sulfur to epoxy-activated 4% cross-linked beaded agarose (C10-spacer)

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Grade For affinity chromatography
Physical Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.