Learn More
ALADDIN SCIENTIFIC CORPORATION Glutathione ReducedGSH Reduced, 10g
Catalog No. 5000082563
Encompass_Preferred
Amino Acid Sequence -Glu-Cys-Gly May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose
Catalog No. 50-000-82563 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. G10542610G
Amino Acid Sequence -Glu-Cys-Gly May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose