Learn More
Arbor Assays Glutathione Reductase Fluorescent Kit (1 Plate)
SDP
Supplier:  Arbor Assays K009F1
Encompass_Preferred
The DetectX Glutathione Reductase (GR) Fluorescent Activity Kit determines GR activity by the amount of GSH generated from the reduction of GSSG with a non-fluorescent molecule, ThioStar, that covalently binds the free thiol group on GSH to yield a highly fluorescent product.
Catalog No. 50-227-8391