Learn More
Invitrogen™ Glutathione Reductase Fluorescent Activity Kit
Description
Includes
96 Well Plates, standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.
- Assay Type: Enzyme Activity kit
- Sample Types: Serum, Plasma, RBCs and Cell Lysates
- Sensitivity: 0.009 mU/mL
- Standard Curve Range: 0.15 mU/mL–5 mU/mL
Specifications
Specifications
| Content And Storage | 96-well plate(s), standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet. Store at –20°C. |
| Immunoassay Kit Format | Assay Activity Kit |
| Product Type | Glutathione Reductase Fluorescent Activity Kit |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| For Use With (Equipment) | Microplate Reader |
| Quantity | 96 tests |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Target Kit Named | Glutathione Reductase |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.