Learn More
Genscript Corporation Glutathione Resin
Catalog No. NC1057345
Encompass
Glutathione Resin (Cat. No. L00206) is an affinity chromatography medium designed for easy, one-step purification of recombinant glutathione S-transferase (GST) fusion proteins and other glutathione binding proteins expressed in E. coli, insect cells and mammalian cells. The recombinant GST fusion proteins can be purified directly from pre-treated cell lysate using Glutathione Resin. It is the excellent choice for high performance purifications. Table 1 lists the main characteristics of Glutathione Resin.
Catalog No. NC1057345 Supplier Genscript Corporation Supplier No. L00206
Glutathione Resin (Cat. No. L00206) is an affinity chromatography medium designed for easy, one-step purification of recombinant glutathione S-transferase (GST) fusion proteins and other glutathione binding proteins expressed in E. coli, insect cells and mammalian cells. The recombinant GST fusion proteins can be purified directly from pre-treated cell lysate using Glutathione Resin. It is the excellent choice for high performance purifications. Table 1 lists the main characteristics of Glutathione Resin.