Learn More
Cusabio Technology LLC glutathione S-transferase pi 1
Catalog No. 5000049179
Encompass_Preferred
This kit detects GSTP1 and is designed for the quantitative measurement of human glutathione S-transferases (GSTpi) in serum plasma cell supernates and tissue homogenates using a sandwich ELISA method It offers a detection range of 1 56 ng/mL to 100 ng/mL with a sensitivity of 0 39 ng/mL ensuring precise quantification of target proteins The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable data for research applications in studying GSTP1 expression and activity across diverse biological samples
Catalog No. 50-000-49179 Supplier Cusabio Technology LLC Supplier No. CSBE09032H
This kit detects GSTP1 and is designed for the quantitative measurement of human glutathione S-transferases (GSTpi) in serum plasma cell supernates and tissue homogenates using a sandwich ELISA method It offers a detection range of 1 56 ng/mL to 100 ng/mL with a sensitivity of 0 39 ng/mL ensuring precise quantification of target proteins The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable data for research applications in studying GSTP1 expression and activity across diverse biological samples