Learn More
Bioworld GLUTATHIONE SEPAROPOREAGA
Supplier:  Bioworld 201820133
Encompass
GLUTATHIONE SEPAROPORE (AGAROSE), HIGH RESOLUTION (HR): Glutathione Separopore HR is a highresolution matrix highly cross-linked 38 m agarose bead (6%)matrix. Glutathione Separopore HRensures the lowest sample dilution and best high-reso...
Catalog No. 50-197-975
Add to Cart
Request a Quote