Learn More
Bioworld GLUTATHIONE SEPAROPOREAGA
Supplier:  Bioworld 201810772
Encompass
GLUTATHIONE SEPAROPORE (AGAROSE) 4B-LYOPHILIZED: Specifications 1 grams make about 10ml gel.; Reduced glutathione (GSH) covalently coupled to Separopore via 10-carbon spacer arm.; For affinity purification of proteins containing glutathi...
Catalog No. 50-197-868