Learn More
Bioworld GLUTATHIONE SEPAROPOREAGA
Catalog No. 50197973
Encompass
GLUTATHIONE SEPAROPORE (AGAROSE), HIGH RESOLUTION (HR): Glutathione Separopore HR is a highresolution matrix highly cross-linked 38 m agarose bead (6%)matrix. Glutathione Separopore HRensures the lowest sample dilution and best high-reso...
Catalog No. 50-197-973 Supplier Bioworld Supplier No. 201820131
GLUTATHIONE SEPAROPORE (AGAROSE), HIGH RESOLUTION (HR): Glutathione Separopore HR is a highresolution matrix highly cross-linked 38 m agarose bead (6%)matrix. Glutathione Separopore HRensures the lowest sample dilution and best high-reso...