Learn More
Bioworld GLUTATHIONE SEPAROPOREAGA
Supplier:  Bioworld 201810501
Encompass
GLUTATHIONE SEPAROPORE (AGAROSE) 4B: Specifications Reduced glutathione (GSH) covalently coupled to Separopore via 10-carbon spacer arm.; For affinity purification of proteins containing glutathione S-transferase (GST) fusion tag.; Suppl...
Catalog No. 50-197-793