Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutathione SepharoseR 4 F
Catalog No. 502908811
Encompass
Glutathione Sepharose(R) 4 Fast Flow 500 mLGlutathione Sepharose(R) 4 Fast Flow is an affinity medium for fast one-step purification of Glutathione S-Transferase (GST) tagged proteins. Provides good flow properties excellent for scale-up.Glutathione Sepharose(R) 4 Fast Flow is used for the purification of GST-tagged proteins from bacterial lysates and other S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function. Sepharose(R) 4 Fast Flow provides good flow properties and is excellent for scaling up.
Catalog No. 50-290-8811 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17513203
Add to Cart
Add to Cart
Request a Quote
Glutathione Sepharose(R) 4 Fast Flow 500 mLGlutathione Sepharose(R) 4 Fast Flow is an affinity medium for fast one-step purification of Glutathione S-Transferase (GST) tagged proteins. Provides good flow properties excellent for scale-up.Glutathione Sepharose(R) 4 Fast Flow is used for the purification of GST-tagged proteins from bacterial lysates and other S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function. Sepharose(R) 4 Fast Flow provides good flow properties and is excellent for scaling up.