Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutathione SepharoseR 4B
Catalog No. 502908660
Encompass
Glutathione Sepharose(R) 4B is designed for high capacity single-step purification of Glutathione S-Transferase (GST) tagged fusion proteins.GST-tagged proteins and other glutathione S-transferases or glutathone-dependent proteins can be purified directly from bacterial lysates using Glutathione Sepharose(R) 4B. Proteins are eluted under mild non-denaturing conditions that preserve protein antigenicity.
Catalog No. 50-290-8660 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17075604
Add to Cart
Add to Cart
Request a Quote
Glutathione Sepharose(R) 4B is designed for high capacity single-step purification of Glutathione S-Transferase (GST) tagged fusion proteins.GST-tagged proteins and other glutathione S-transferases or glutathone-dependent proteins can be purified directly from bacterial lysates using Glutathione Sepharose(R) 4B. Proteins are eluted under mild non-denaturing conditions that preserve protein antigenicity.