Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutathione SepharoseR 4B
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17075601
Encompass
Glutathione Sepharose(R) 4B is designed for high capacity single-step purification of Glutathione S-Transferase (GST) tagged fusion proteins.GST-tagged proteins and other glutathione S-transferases or glutathone-dependent proteins can be purified directly from bacterial lysates using Glutathione Sepharose(R) 4B. Proteins are eluted under mild non-denaturing conditions that preserve protein antigenicity.
Catalog No. 50-290-4830