Learn More
Medchemexpress LLC Glutaurine TFA 5mg | 5MG
SDP
Catalog No. 5000241022
Encompass_Preferred
Glutaurine (Litoralon) TFA containing glutamine and taurine residues is an orally active hormone of the parathyroid Glutaurine (Litoralon) TFA as a hormone is isolated from parathyroid gland oxyphil cells Glutaurine (Litoralon) TFA can be used for the research of antiepileptic and anti-amnesia[1 [2 [3 [4
Catalog No. 50-002-41022 Supplier Medchemexpress LLC Supplier No. HY106608A5MG
Glutaurine (Litoralon) TFA containing glutamine and taurine residues is an orally active hormone of the parathyroid Glutaurine (Litoralon) TFA as a hormone is isolated from parathyroid gland oxyphil cells Glutaurine (Litoralon) TFA can be used for the research of antiepileptic and anti-amnesia[1 [2 [3 [4