Learn More
Polysciences, Inc. Glutaraldehyde; Polysciences; EM Grade; 25%; Stable; Clear colorless liquid acrid odor; Stored at 4 deg. C; Refractive index: 1.375; Density: 1.062; CAS: [111-30-8]; Size: 10 x 10mL
SDP
Catalog No. NC9861069
Encompass
Glutaraldehyde; Polysciences; EM Grade; 25%; Stable; Clear colorless liquid acrid odor; Stored at 4 deg. C; Refractive index: 1.375; Density: 1.062; CAS: [111-30-8]; Size: 10 x 10mL
Catalog No. NC9861069 Supplier Polysciences, Inc. Supplier No. 0190910
Glutaraldehyde; Polysciences; EM Grade; 25%; Stable; Clear colorless liquid acrid odor; Stored at 4 deg. C; Refractive index: 1.375; Density: 1.062; CAS: [111-30-8]; Size: 10 x 10mL