Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Glutaraldehyde 2.5% Paraformaldehyde 1% in Phosphate Buffer, pH 7.4 50 ML
SDP

Catalog No. 501901192
Encompass_Preferred

Glutaraldehyde 2.5% Paraformaldehyde 1% in Phosphate Buffer.
pH: 7.4

Catalog No. 50-190-1192 Supplier Electron Microscopy Sciences Supplier No. 1653707
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutaraldehyde 2.5% Paraformaldehyde 1% in Phosphate Buffer.
pH: 7.4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.