Learn More
MSC Honeywell North Chemical Resistant Gloves, Butyl, 16 mil, 1 Pair
Catalog No. 502370309
Encompass
Honeywell North Butyl Unsupported Chemical Resistant Gloves provide excellent protection against a wide range of chemicals, including vapors, gases, ketones, and esters. Constructed from butyl rubber, these gloves maintain flexibility even in cold environments, reducing hand fatigue and enhancing user comfort during tasks such as chemical handling, laboratory applications, and Hazmat operations.
  • Made from butyl material for superior chemical resistance
  • Large glove size, numeric size 9
  • Provides 16 mil thickness for robust protection
  • Overall length of 11 in
  • Smooth finish provides enhanced tactile sensitivity
  • Rolled cuff style for easy donning and doffing
  • Unlined interior for direct contact and dexterity
  • Unsupported design ideal for light-to-medium chemical applications
  • Retains flexibility in low temperatures, minimizing hand fatigue
Catalog No. 50-237-0309 Supplier MSC Supplier No. 01904119
Honeywell North Butyl Unsupported Chemical Resistant Gloves provide excellent protection against a wide range of chemicals, including vapors, gases, ketones, and esters. Constructed from butyl rubber, these gloves maintain flexibility even in cold environments, reducing hand fatigue and enhancing user comfort during tasks such as chemical handling, laboratory applications, and Hazmat operations.
  • Made from butyl material for superior chemical resistance
  • Large glove size, numeric size 9
  • Provides 16 mil thickness for robust protection
  • Overall length of 11 in
  • Smooth finish provides enhanced tactile sensitivity
  • Rolled cuff style for easy donning and doffing
  • Unlined interior for direct contact and dexterity
  • Unsupported design ideal for light-to-medium chemical applications
  • Retains flexibility in low temperatures, minimizing hand fatigue