Learn More
MSC North Chemical Resistant Gloves, Butyl, 16 mil, X-Large, 1 Pair
Catalog No. 502370355
Encompass
These unsupported butyl gloves are engineered for demanding tasks requiring chemical resistance and dexterity. They provide superior protection against vapors, gases, ketones, esters, ozone, and other oxidizing chemicals, making them ideal for environments with highly toxic chemical hazards.
  • Exceptional permeation resistance to vapors and gases
  • High resistance to ketones (MEK, MIBK, acetone), esters, ozone, alcohols, and acids
  • Soft and flexible design reduces hand fatigue, enhancing comfort
  • Suitable for highly toxic chemical hazards and glove box operations
  • Constructed from 16 mil butyl material
  • Overall length is 11 in with a rolled cuff
Catalog No. 50-237-0355 Supplier MSC Supplier No. 01904127
These unsupported butyl gloves are engineered for demanding tasks requiring chemical resistance and dexterity. They provide superior protection against vapors, gases, ketones, esters, ozone, and other oxidizing chemicals, making them ideal for environments with highly toxic chemical hazards.
  • Exceptional permeation resistance to vapors and gases
  • High resistance to ketones (MEK, MIBK, acetone), esters, ozone, alcohols, and acids
  • Soft and flexible design reduces hand fatigue, enhancing comfort
  • Suitable for highly toxic chemical hazards and glove box operations
  • Constructed from 16 mil butyl material
  • Overall length is 11 in with a rolled cuff