Learn More
ORS Nasco GLV 15IN 20ML CLNRM SZ9 10PRPK
Supplier:  ORS Nasco 068AK1815/O/9
Encompass_Preferred
Honeywell part# AK1815/O/9. Honeywell AK Latex Cleanroom Gloves Size: 9 Certifications/Compliance: IEST-RP-CC005.3ASTM F739 Certifications/Compliance: EN388 EN374-3 Length: 15 in. Length: 38 cm Cleanroom Class: Class 100/ISO 5 Exterior Finish: Partially Textured Laboratory/Cleanroom Use: Yes Latex Free: No
Catalog No. 50-255-1650