Learn More
ORS Nasco GLV 4H CHEMRES SZ11/12 10PR/PK
Catalog No. 502551619
Encompass_Preferred
Honeywell part# SSG/11. Honeywell SilverShield SSG Gloves Length: 14.5 in. 37 cm Size: 11 Glove Cuff Style: Straight Latex Free: Yes Polyethylene Ethylene Vinyl Alcohol Thickness: 2.7 mil 0.06 mm Certifications/Compliance: OSHA 29 CFR 1915.157 EN 420 EN 374-3 EN388 Standard: EN 388 Grip Design: Smooth
Catalog No. 50-255-1619 Supplier ORS Nasco Supplier No. 068SSG/11
Honeywell part# SSG/11. Honeywell SilverShield SSG Gloves Length: 14.5 in. 37 cm Size: 11 Glove Cuff Style: Straight Latex Free: Yes Polyethylene Ethylene Vinyl Alcohol Thickness: 2.7 mil 0.06 mm Certifications/Compliance: OSHA 29 CFR 1915.157 EN 420 EN 374-3 EN388 Standard: EN 388 Grip Design: Smooth