Learn More
MSC Honeywell North Butyl Gloves, 14 mil, Rough, Medium, 1 Pair
Catalog No. 502370351
Encompass
These unsupported butyl gloves are designed for chemical resistance, featuring a 14 mil thickness and a rough textured grip. Sized Medium and sold as a pair, they are black and FDA approved for various applications.
  • Soft flexible feel
  • Curved hand and finger design
  • Sanitized for hygiene
  • Seamless construction
  • FDA approved
  • Excellent permeation resistance to gas and water vapor
  • Highly resistant to ketones and esters
  • Economical for light-to-medium chemical tasks
  • Provides increased dexterity for intricate work
Catalog No. 50-237-0351 Supplier MSC Supplier No. 42104257
These unsupported butyl gloves are designed for chemical resistance, featuring a 14 mil thickness and a rough textured grip. Sized Medium and sold as a pair, they are black and FDA approved for various applications.
  • Soft flexible feel
  • Curved hand and finger design
  • Sanitized for hygiene
  • Seamless construction
  • FDA approved
  • Excellent permeation resistance to gas and water vapor
  • Highly resistant to ketones and esters
  • Economical for light-to-medium chemical tasks
  • Provides increased dexterity for intricate work