Learn More
MSC Honeywell North Butyl Gloves, Rough, 14 mil, XL, 1 pair
Catalog No. 502370354
Encompass
These unsupported Butyl gloves offer exceptional resistance to gas and water vapor, making them ideal for applications involving ketones (such as MEK, MIBL, acetone) and esters (including tricresyl phosphate, amyl acetate, and ethyl acetate).
  • Made from butyl material with 14 mil thickness
  • Unsupported and unlined construction
  • X-Large size with 14 in overall length
  • Rough textured grip for enhanced handling
  • Beaded cuff for easy donning and doffing
  • Provides resistance to abrasion, cut, puncture, and tear
  • FDA approved for safe use
Catalog No. 50-237-0354 Supplier MSC Supplier No. 51358687
These unsupported Butyl gloves offer exceptional resistance to gas and water vapor, making them ideal for applications involving ketones (such as MEK, MIBL, acetone) and esters (including tricresyl phosphate, amyl acetate, and ethyl acetate).
  • Made from butyl material with 14 mil thickness
  • Unsupported and unlined construction
  • X-Large size with 14 in overall length
  • Rough textured grip for enhanced handling
  • Beaded cuff for easy donning and doffing
  • Provides resistance to abrasion, cut, puncture, and tear
  • FDA approved for safe use