Learn More
MSC North Chemical Resistant Gloves, Butyl, Large, 17 mil, 1 Pair
Catalog No. 502370297
Encompass
These chemical resistant gloves are made from butyl, designed for light-to-medium chemical applications requiring dexterity. They feature an unsupported construction with a Grip-Saf grip.
  • Made from butyl material for chemical resistance
  • Large size (numeric size 9)
  • Provides 17 mil thickness for protection
  • Unsupported design for economy and dexterity
  • Textured finish for improved grip
  • Black color
  • Rolled cuff style
  • Unlined for enhanced tactile sensitivity
  • Features 14.00 in overall length
Catalog No. 50-237-0297 Supplier MSC Supplier No. 75272914
These chemical resistant gloves are made from butyl, designed for light-to-medium chemical applications requiring dexterity. They feature an unsupported construction with a Grip-Saf grip.
  • Made from butyl material for chemical resistance
  • Large size (numeric size 9)
  • Provides 17 mil thickness for protection
  • Unsupported design for economy and dexterity
  • Textured finish for improved grip
  • Black color
  • Rolled cuff style
  • Unlined for enhanced tactile sensitivity
  • Features 14.00 in overall length