Learn More
MSC Honeywell North Nitrile Gloves, Medium, Powder-Free, Cleanroom ISO 5, 100/Pack
Catalog No. 502370310
Encompass
North Chemsoft CE disposable nitrile gloves are designed for cleanroom environments, offering a combination of comfort and protection. These ambidextrous, powder-free gloves feature a grain (fingertips) grip and are supplied in cleanroom-compatible packaging.
  • Packaged and double-bagged with cleanroom compatible materials
  • Surface resistivity of 10^10 ohms/sq
  • Low-modulus formulation
  • Extended and rolled cuff for secure fit
  • Antistatic properties
  • Provides Type B chemical protection against n-Heptane, Sodium Hydroxide 40%, and Formaldehyde 37%
  • Protects against bacteria and fungi per EN ISO 374-5
  • Nominal palm thickness of 4 mil
  • ISO 5 cleanroom compatible
Catalog No. 50-237-0310 Supplier MSC Supplier No. 01293927
North Chemsoft CE disposable nitrile gloves are designed for cleanroom environments, offering a combination of comfort and protection. These ambidextrous, powder-free gloves feature a grain (fingertips) grip and are supplied in cleanroom-compatible packaging.
  • Packaged and double-bagged with cleanroom compatible materials
  • Surface resistivity of 10^10 ohms/sq
  • Low-modulus formulation
  • Extended and rolled cuff for secure fit
  • Antistatic properties
  • Provides Type B chemical protection against n-Heptane, Sodium Hydroxide 40%, and Formaldehyde 37%
  • Protects against bacteria and fungi per EN ISO 374-5
  • Nominal palm thickness of 4 mil
  • ISO 5 cleanroom compatible