Learn More
MSC Honeywell Disposable Gloves, Nitrile, Small, 4 mil, Cleanroom, Powder-Free, 100/Pack
Catalog No. 502370312
Encompass
Honeywell North Chemsoft disposable nitrile gloves are engineered for cleanroom environments, offering reliable hand protection with a focus on cleanliness and user comfort.
  • Made from nitrile material for chemical resistance
  • Features 4 mil nominal palm thickness for tactile sensitivity
  • Cleanroom grade, ISO 5 rated
  • Powder-free and ambidextrous design
  • Features grain (fingertips) grip for enhanced dexterity
  • Extended, rolled cuff type
  • Surface resistivity 10^10 Ohms/sq
  • Low-modulus formulation for reduced hand fatigue
Catalog No. 50-237-0312 Supplier MSC Supplier No. 01293919
Honeywell North Chemsoft disposable nitrile gloves are engineered for cleanroom environments, offering reliable hand protection with a focus on cleanliness and user comfort.
  • Made from nitrile material for chemical resistance
  • Features 4 mil nominal palm thickness for tactile sensitivity
  • Cleanroom grade, ISO 5 rated
  • Powder-free and ambidextrous design
  • Features grain (fingertips) grip for enhanced dexterity
  • Extended, rolled cuff type
  • Surface resistivity 10^10 Ohms/sq
  • Low-modulus formulation for reduced hand fatigue